2-Chloro-1,3-dimethylimidazolinium hexafluorophosphate ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
CIP, 2-Chloro-1,3-dimethyl-4,5-dihydro-1H-imidazol-3-ium hexafluorophosphate(V)
C1651-25G
C1651-5G
TCIAC1651-25GEA
977
DKK
TCIAC1651-25G
TCIAC1651-5G
2-Chloro-1,3-dimethylimidazolinium hexafluorophosphate ≥98.0% (by HPLC, titration analysis)
2-Chloro-1,3-dimethylimidazolinium hexafluorophosphate
Formel:
C₅H₁₀ClF₆N₂P |
MDL Number:
MFCD00191914 CAS-nummer: 101385-69-7 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 233.0 to 237.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...