Chloromethyl polystyrene resin 0,8 - 1,3 mmol/g, cross-linked with 1% DVB (200-400 mesh)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Chloromethylated styrene-divinylbenzene copolymer, Merrifield Resin
C1642-25G C1642-5G
TCIAC1642-25GEA 2700 DKK
TCIAC1642-25G TCIAC1642-5G
Chloromethyl polystyrene resin 0,8 - 1,3 mmol/g, cross-linked with 1% DVB (200-400 mesh)
Chloromethyl polystyrene resin
MDL Number: MFCD00146406
CAS-nummer: 55844-94-5
Bestil nu
Specifikationer Testresultater
Loading 0.8 to 1.3 mmol/g