1-(4-Chlorobenzhydryl)piperazine ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-(4-Chlorodiphenylmethyl)piperazine, 1-((4-Chlorophenyl)(phenyl)methyl)piperazine

C1611-5G C1611-1G
TCIAC1611-5GEA 764 DKK
TCIAC1611-5G TCIAC1611-1G
1-(4-Chlorobenzhydryl)piperazine ≥98.0% (by GC)
1-(4-Chlorobenzhydryl)piperazine
Formel: C₁₇H₁₉ClN₂
MDL Number: MFCD00005965
CAS-nummer: 303-26-4
EINECS: 206-137-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=578be2d8f88b6b94bb408ce82a4e61b2470ac8a8&vid=578be2d8f88b6b94bb408ce82a4e61b2470ac8a8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR