Cycloheptanone oxime ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
C1543-25G
C1543-5G
TCIAC1543-25GEA
1650
DKK
TCIAC1543-25G
TCIAC1543-5G
Cycloheptanone oxime ≥98.0% (by GC)
Cycloheptanone oxime
Formel:
C₇H₁₃NO Kogepunkt: 152°C |
MDL Number:
MFCD00046508 CAS-nummer: 2158-31-8 Merck Index: 14,02722 |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...