N-(2-Cyanoethyl)glycine ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: N-(2-Cyanoethyl)aminoacetic Acid

C1525-25G C1525-5G
TCIAC1525-25GEA 731 DKK
TCIAC1525-25G TCIAC1525-5G
N-(2-Cyanoethyl)glycine ≥98.0% (by titrimetric analysis)
N-(2-Cyanoethyl)glycine
Formel: C₅H₈N₂O₂
MDL Number: MFCD00066903
CAS-nummer: 3088-42-4
EINECS: 221-418-1

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR