N-Carbobenzoxy-1,4-diaminobutane hydrochloride ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: N-Z-1,4-Butanediamine hydrochloride, N-Cbz-1,4-diaminobutane Hydrochloride, Benzyl N-(4-Aminobutyl)carbamate Hydrochloride, Benzyl (4-aminobutyl)carbamate hydrochloride, N-(4-Aminobutyl)carbamic Acid Benzyl Ester Hydrochloride

C1519-1G C1519-5G
TCIAC1519-1GEA 493 DKK
TCIAC1519-1G TCIAC1519-5G
N-Carbobenzoxy-1,4-diaminobutane hydrochloride ≥97.0% (by titrimetric analysis)
N-Carbobenzoxy-1,4-diaminobutane hydrochloride
Formel: C₁₂H₁₈N₂O₂
Molekylvægt: 258.75 g/mol
MDL Number: MFCD00270149
CAS-nummer: 18807-73-3

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR