N-Carbobenzoxy-1,3-diaminopropane hydrochloride ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: N-Benzyloxycarbonyl-1,3-propanediamine hydrochloride, N-Z-1,3-Propanediamine hydrochloride, N-Benzyloxycarbonyl-1,3-diaminopropane hydrochloride, N-Cbz-1,3-diaminopropane Hydrochloride, Benzyl N-(3-Aminopropyl)carbamate Hydrochloride, N-(3-Aminopropyl)carbamic Acid Benzyl Ester Hydrochloride, Benzyl (3-aminopropyl)carbamate hydrochloride

C1512-1G C1512-5G
TCIAC1512-1GEA 312 DKK
TCIAC1512-1G TCIAC1512-5G
N-Carbobenzoxy-1,3-diaminopropane hydrochloride ≥98.0% (by titrimetric analysis)
N-Carbobenzoxy-1,3-diaminopropane hydrochloride
Formel: C₁₁H₁₆N₂O₂
MDL Number: MFCD00270153
CAS-nummer: 17400-34-9

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 183.0 to 189.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR