Ethephon(2-chloroethylphosphonic acid) ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Chloroethylphosphonic acid, Ethephon, (2-Chloroethyl)phosphonic acid

C1456-1G C1456-5G
TCIAC1456-1GEA 714 DKK
TCIAC1456-1G TCIAC1456-5G
Ethephon(2-chloroethylphosphonic acid) ≥98.0% (by titrimetric analysis)
Ethephon(2-chloroethylphosphonic acid)
Formel: C₂H₆ClO₃P
Kogepunkt: 265°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00002142
CAS-nummer: 16672-87-0
EINECS: 240-718-3
UN nummer: 2923
ADR: 8 II
Merck Index: 14,03732

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 73.0 to 78.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR