N-(Carbobenzoxy)aminoacetonitrile ≥95.0% (by total nitrogen basis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Benzyl (cyanomethyl)carbamate, Cyanomethylcarbamic Acid Benzyl Ester, Benzyl Cyanomethylcarbamate, N-Cbz-aminoacetonitrile
C1439-25G
C1439-5G
TCIAC1439-25GEA
5010
DKK
TCIAC1439-25G
TCIAC1439-5G
N-(Carbobenzoxy)aminoacetonitrile ≥95.0% (by total nitrogen basis)
N-(Carbobenzoxy)aminoacetonitrile
Formel:
C₁₀H₁₀N₂O₂ |
MDL Number:
MFCD00033175 CAS-nummer: 3589-41-1 |
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (with Total Nitrogen) | min. 95.0 % |
Melting point | 58.0 to 61.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...