1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium hexafluorophosphate ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-(Chloro(pyrrolidin-1-yl)methylene)pyrrolidin-1-ium hexafluorophosphate(V), Chloro-N,N,N',N'-bis(tetramethylene)formamidinium Hexafluorophosphate, PyClU

C1379-25G C1379-5G
TCIAC1379-25GEA 1260 DKK
TCIAC1379-25G TCIAC1379-5G
1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium hexafluorophosphate ≥97.0% (by titrimetric analysis)
1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium hexafluorophosphate
Formel: C₉H₁₆ClF₆N₂P
MDL Number: MFCD00191333
CAS-nummer: 135540-11-3

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (Argentmetric Titration) min. 97.0 %
Melting point 150.0 to 155.0 deg-C
Solubility in Acetonitrile almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR