Hexanal 2,4-Dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Capronaldehyde 2,4-Dinitrophenylhydrazone

C1330-1G C1330-5G
TCIAC1330-1GEA 189 DKK
TCIAC1330-1G TCIAC1330-5G
Hexanal 2,4-Dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)
Hexanal 2,4-Dinitrophenylhydrazone
Formel: C₁₂H₁₆N₄O₄
Molekylvægt: 280.28 g/mol
MDL Number: MFCD00191624
CAS-nummer: 1527-97-5
UN nummer: 1325
ADR: 4.1 III

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 106.0 to 110.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR