Cumylamine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C1293-25ML C1293-5ML
TCIAC1293-25MLEA 2170 DKK
TCIAC1293-25ML TCIAC1293-5ML
Cumylamine ≥98.0% (by GC, titration analysis)
Cumylamine
Formel: C₉H₁₃N
Kogepunkt: 197°C
MDL Number: MFCD00134680
CAS-nummer: 585-32-0

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR