Triethyl orthochloroacetate ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Orthochloroacetic Acid Triethyl Ester, 2-Chloro-1,1,1-triethoxyethane
C1288-1ML
C1288-5ML
TCIAC1288-1MLEA
583
DKK
TCIAC1288-1ML
TCIAC1288-5ML
Triethyl orthochloroacetate ≥95.0% (by GC)
Triethyl orthochloroacetate
Formel:
C₈H₁₇ClO₃ Molekylvægt: 196.67 g/mol Kogepunkt: 91°C |
MDL Number:
MFCD00142889 CAS-nummer: 51076-95-0 |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...