2-(Chloromethyl)-3,5-dioxahex-1-ene ≥96.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-Chloro-2-(methoxymethoxy)-1-propene
C1198-5G
C1198-1G
TCIAC1198-5GEA
1700
DKK
TCIAC1198-5G
TCIAC1198-1G
2-(Chloromethyl)-3,5-dioxahex-1-ene ≥96.0% (by GC)
2-(Chloromethyl)-3,5-dioxahex-1-ene
Formel:
C₅H₉ClO₂ Molekylvægt: 136.58 g/mol Kogepunkt: 63°C |
MDL Number:
MFCD00130115 CAS-nummer: 105104-40-3 UN nummer: 1993 ADR: 3 III |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...