1-Chloro-2-(chloromethyl)-3,5-dioxahexane ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1,3-Dichloro-2-(methoxymethoxy)propane, 1,1-Bis(chloromethyl)-2,4-dioxapentane

C1197-25G C1197-5G
TCIAC1197-25GEA 1190 DKK
TCIAC1197-25G TCIAC1197-5G
1-Chloro-2-(chloromethyl)-3,5-dioxahexane ≥95.0% (by GC)
1-Chloro-2-(chloromethyl)-3,5-dioxahexane
Formel: C₅H₁₀Cl₂O₂
Molekylvægt: 173.03 g/mol
Kogepunkt: 95°C
MDL Number: MFCD00130114
CAS-nummer: 70905-45-2

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR