sec-Butyl crotonate ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Crotonic Acid sec-Butyl Ester
C1184-25ML
C1184-250ML
TCIAC1184-25MLEA
394
DKK
TCIAC1184-25ML
TCIAC1184-250ML
sec-Butyl crotonate ≥97.0% (by GC)
sec-Butyl crotonate
Formel:
C₈H₁₄O₂ Molekylvægt: 142.20 g/mol Kogepunkt: 166°C |
MDL Number:
MFCD00059054 CAS-nummer: 10371-45-6 UN nummer: 3272 ADR: 3 III |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...