1-Cyclohexyl-2-pyrrolidone ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

C0963-25ML C0963-500ML
TCIAC0963-25MLEA 140 DKK
TCIAC0963-25ML TCIAC0963-500ML
1-Cyclohexyl-2-pyrrolidone ≥99.0% (by GC)
1-Cyclohexyl-2-pyrrolidone
Formel: C₁₀H₁₇NO
Kogepunkt: 154°C
Flammepunkt: 141°C
MDL Number: MFCD00003191
CAS-nummer: 6837-24-7
EINECS: 229-919-7
UN nummer: 2810
ADR: 6.1,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Orange to Green clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR