Cyclopentylmalonic acid ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
C0945-25G
C0945-5G
TCIAC0945-25GEA
2690
DKK
TCIAC0945-25G
TCIAC0945-5G
Cyclopentylmalonic acid ≥98.0% (by titrimetric analysis)
Cyclopentylmalonic acid
Formel:
C₈H₁₂O₄ |
MDL Number:
MFCD00059515 CAS-nummer: 5660-81-1 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 160.0 to 165.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...