Vinyl-n-octanoate ≥99.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: n-Octanoic Acid Vinyl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

C0896-25ML C0896-500ML
TCIAC0896-25MLEA 189 DKK
TCIAC0896-25ML TCIAC0896-500ML
Vinyl-n-octanoate ≥99.0% (by GC) stabilised
Vinyl-n-octanoate
Formel: C₁₀H₁₈O₂
Molekylvægt: 170.25 g/mol
Kogepunkt: 135°C
Flammepunkt: 75°C
MDL Number: MFCD00048915
CAS-nummer: 818-44-0
EINECS: 212-452-8

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR