Cyclohexanecarboxaldehyde ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C0880-25ML C0880-100ML
TCIAC0880-25MLEA 411 DKK
TCIAC0880-25ML TCIAC0880-100ML
Cyclohexanecarboxaldehyde ≥98.0% (by GC)
Cyclohexanecarboxaldehyde
Formel: C₇H₁₂O
Molekylvægt: 112.17 g/mol
Kogepunkt: 159°C
Flammepunkt: 40°C
MDL Number: MFCD00001457
CAS-nummer: 2043-61-0
EINECS: 218-057-7
UN nummer: 1989
ADR: 3 III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR