p-Tolyl crotonate >55.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Crotonic Acid p-Cresyl Ester, Crotonic Acid p-Tolyl Ester, p-Cresyl Crotonate

C0866-25ML C0866-500ML
TCIAC0866-25MLEA 222 DKK
TCIAC0866-25ML TCIAC0866-500ML
p-Tolyl crotonate >55.0% (by GC)
p-Tolyl crotonate
Formel: C₁₁H₁₂O₂
Molekylvægt: 176.22 g/mol
Kogepunkt: 158°C
MDL Number: MFCD00059385
CAS-nummer: 41873-74-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec7d5f13fdf95c7ec8efa8882cd610058a9b2f8d&vid=ec7d5f13fdf95c7ec8efa8882cd610058a9b2f8d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 55.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR