3-Carboxy-1-(4-sulphophenyl)-5-pyrazolone sodium salt ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

C0845-25G C0845-500G
TCIAC0845-25GEA 312 DKK
TCIAC0845-25G TCIAC0845-500G
3-Carboxy-1-(4-sulphophenyl)-5-pyrazolone sodium salt ≥98.0% (by HPLC, titration analysis)
3-Carboxy-1-(4-sulphophenyl)-5-pyrazolone sodium salt
Formel: C₁₀H₈N₂O₆S
MDL Number: MFCD00059720
CAS-nummer: 52126-51-9
EINECS: 257-677-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76fe4f5c53215655162d2873c210468b27886c17&vid=96903bf3ea5006b3fa0dadac6b17f9b4eed06fee&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR