N-Carbobenzoxy-L-methionine ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: N-Carbobenzyloxy-L-methionine, Z-Met-OH, N-Cbz-L-methionine, Cbz-Met-OH

C0737-25G C0737-5G
TCIAC0737-25GEA 526 DKK
TCIAC0737-25G TCIAC0737-5G
N-Carbobenzoxy-L-methionine ≥98.0% (by titrimetric analysis)
N-Carbobenzoxy-L-methionine
Formel: C₁₃H₁₇NO₄S
MDL Number: MFCD00065124
CAS-nummer: 1152-62-1
EINECS: 214-570-5

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 67.0 to 70.0 deg-C
Specific rotation [a]20/D -25.0 to -27.0 deg (C=1, MeOH)
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR