Phosphonitrilic chloride trimer ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

C0584-25G C0584-250G
TCIAC0584-25GEA 378 DKK
TCIAC0584-25G TCIAC0584-250G
Phosphonitrilic chloride trimer ≥98.0% (by GC)
Phosphonitrilic chloride trimer
Formel: Cl₆N₃P₃
MDL Number: MFCD00006474
CAS-nummer: 940-71-6
EINECS: 213-376-8
UN nummer: 1759
ADR: 8,III

Bestil nu

Specifikationer Testresultater

Appearance White powder to crystal
Purity (GC) min. 98.0 %
Melting point 110.0 to 114.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR