Curcumin (natural)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione
C0434-1G
C0434-25G
TCIAC0434-1GEA
156
DKK
TCIAC0434-1G
TCIAC0434-25G
Curcumin (natural)
Curcumin
Formel:
C₂₁H₂₀O₆ Molekylvægt: 368.39 g/mol |
MDL Number:
MFCD00008365 CAS-nummer: 458-37-7 EINECS: 207-280-5 Merck Index: 14,02673 |
|
Specifikationer Testresultater
Appearance | Light yellow to Brown powder to crystal |
Absorbance | min. 0.340 (2.5mg/L, EtOH, 428.0 to 432.0 nm) |
Sensitiveness | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...