Choline dihydrogen citrate ≥99.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Choline dihydrogencitrate salt, 2-Hydroxy-N,N,N-trimethylethan-1-aminium 3,4-dicarboxy-3-hydroxybutanoate
C0331-25G
C0331-500G
TCIAC0331-25GEA
132
DKK
TCIAC0331-25G
TCIAC0331-500G
Choline dihydrogen citrate ≥99.0% (by titrimetric analysis)
Choline dihydrogen citrate
Formel:
C₁₁H₂₁NO₈ Molekylvægt: 295.29 g/mol |
MDL Number:
MFCD00038734 CAS-nummer: 77-91-8 EINECS: 201-068-6 Merck Index: 14,02207 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 99.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...