Chlorophenol red sodium salt

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C0247-1G C0247-5G
TCIAC0247-1GEA 132 DKK
TCIAC0247-1G TCIAC0247-5G
Chlorophenol red sodium salt
Chlorophenol red sodium salt
Formel: C₁₉H₁₂Cl₂O₅S
Molekylvægt: 445.24 g/mol
MDL Number: MFCD00151199
CAS-nummer: 123333-64-2

Bestil nu

Specifikationer Testresultater

Appearance Red to Dark blue to Black powder to crystal
Drying loss max. 3.0 %
Transition interval (pH) (yellow)5.0 to 6.6 (red purple)

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR