3-Chlorobutyric acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
C0147-10ML
C0147-25ML
TCIAC0147-10MLEA
1510
DKK
TCIAC0147-10ML
TCIAC0147-25ML
3-Chlorobutyric acid ≥98.0% (by GC, titration analysis)
3-Chlorobutyric acid
Formel:
C₄H₇ClO₂ Molekylvægt: 122.55 g/mol Kogepunkt: 112°C |
MDL Number:
MFCD00020496 CAS-nummer: 1951-12-8 EINECS: 217-771-6 UN nummer: 3265 ADR: 8 III |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...