Carboxymethylcellulosenatriumsalt n=∼ 500

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Carboxymethyl cellulose, sodium salt, Cellulose gum, Cellulose, carboxymethyl ether, sodium salt, Sodium carboxymethyl cellulose (Viscosity 800mpa.s), Sodium carboxymethyl cellulose, CMC

C0045-25G C0045-500G
TCIAC0045-25GEA 148 DKK
TCIAC0045-25G TCIAC0045-500G
Carboxymethylcellulosenatriumsalt n=∼ 500
Carboxymethylcellulosenatriumsalt
Formel: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
MDL Number: MFCD00081472
CAS-nummer: 9004-32-4
Merck Index: 14,01829

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Content (Na,Drying substance) 6.0 to 8.5 %
Etherification value (as Drying substance) 0.5 to 0.8
Drying loss max. 10.0 %
Viscosity 500.0 to 900.0 mPa-s (2 %, H2O, 25 deg-C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR