Nε-carbobenzoxy-L-lysine ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Nε-Cbz-L-lysine, (S)-2-Amino-6-(((benzyloxy)carbonyl)amino)hexanoic acid, H-Lys(Z)-OH

C0034-25G C0034-5G
TCIAC0034-25GEA 657 DKK
TCIAC0034-25G TCIAC0034-5G
Nε-carbobenzoxy-L-lysine ≥98.0% (by titrimetric analysis)
Nepsilon-carbobenzoxy-L-lysine
Formel: C₁₄H₂₀N₂O₄
Molekylvægt: 280.32 g/mol
MDL Number: MFCD00002638
CAS-nummer: 1155-64-2
EINECS: 214-585-7

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +15.0 to +17.0 deg (C=1.6, 2mol/L HCl)
Solubility in hot HCl almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR