sec-Butyl sulphide ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B6136-25ML
B6136-5ML
TCIAB6136-25MLEA
1020
DKK
TCIAB6136-25ML
TCIAB6136-5ML
sec-Butyl sulphide ≥98.0% (by GC)
sec-Butyl sulphide
Formel:
C₈H₁₈S Kogepunkt: 165°C Flammepunkt: 39°C |
MDL Number:
MFCD00009332 CAS-nummer: 626-26-6 EINECS: 210-938-4 UN nummer: 1993 ADR: 3,III |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...