N-(4-Bromophenyl)-9,9-dimethyl-9H-fluoren-2-amine ≥95.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B6133-1G B6133-5G
TCIAB6133-1GEA 550 DKK
TCIAB6133-1G TCIAB6133-5G
N-(4-Bromophenyl)-9,9-dimethyl-9H-fluoren-2-amine ≥95.0% (by HPLC, total nitrogen)
N-(4-Bromophenyl)-9,9-dimethyl-9H-fluoren-2-amine
Formel: C₂₁H₁₈BrN
MDL Number: MFCD32263467
CAS-nummer: 1644059-09-5

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Melting point 122.0 to 126.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR