Bis(4-pyridyl)amine ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B6124-1G
B6124-250MG
TCIAB6124-1GEA
1400
DKK
TCIAB6124-1G
TCIAB6124-250MG
Bis(4-pyridyl)amine ≥98.0% (by GC, titration analysis)
Formel:
C₁₀H₉N₃ |
MDL Number:
MFCD00829434 CAS-nummer: 1915-42-0 |
Specifikationer Testresultater
Appearance | White to Green to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...