2-(2-Bromophenyl)naphthalene ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B6100-1G B6100-5G
TCIAB6100-1GEA 862 DKK
TCIAB6100-1G TCIAB6100-5G
2-(2-Bromophenyl)naphthalene ≥98.0% (by GC)
2-(2-Bromophenyl)naphthalene
Formel: C₁₆H₁₁Br
MDL Number: MFCD18415513
CAS-nummer: 22082-97-9

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 80.0 to 84.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR