tert-Butyl-3-formylpyrrolidine-1-carboxylate ≥96.0% (by GC)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: (+/-)-3-Formylpyrrolidine-1-carboxylic acid tert-Butyl ester, (+/-)-1-Boc-pyrrolidine-3-carboxaldehyde, (+/-)-1-(tert-Butoxycarbonyl)-3-formylpyrrolidine, (+/-)-1-Boc-3-formylpyrrolidine, tert-Butyl 3-formylpyrrolidine-1-carboxylate, (+/-)-tert-Butyl 3-Formylpyrrolidine-1-carboxylate, (+/-)-1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxaldehyde
B6091-1G B6091-5G
TCIAB6091-1GEA 468 DKK
TCIAB6091-1G TCIAB6091-5G
tert-Butyl-3-formylpyrrolidine-1-carboxylate ≥96.0% (by GC)
tert-Butyl-3-formylpyrrolidine-1-carboxylate
Formel: C₁₀H₁₇NO₃
Molekylvægt: 199.2500 g/mol
MDL Number: MFCD03791257
CAS-nummer: 59379-02-1
Bestil nu
Specifikationer Testresultater
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %