Boc-Amino-PEG3-Amine ≥96.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
tert-Butyl (2-(2-(2-(2-aminoethoxy)ethoxy)ethoxy)ethyl)carbamate, NH2-PEG3-C2-NH-Boc
B6080-1G
B6080-250MG
TCIAB6080-1GEA
2520
DKK
TCIAB6080-1G
TCIAB6080-250MG
Boc-Amino-PEG3-Amine ≥96.0% (by GC, titration analysis)
Boc-Amino-PEG3-Amine
Formel:
C₁₃H₂₈N₂O₅ Opbevaringstemperatur: Fryser |
MDL Number:
MFCD16619220 CAS-nummer: 101187-40-0 |
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid to cloudy liquid |
Purity (GC) | min. 96.0 % |
Purity (Neutralization titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...