3-(Benzyloxy)cyclobutanone ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B6029-1G
B6029-200MG
TCIAB6029-1GEA
624
DKK
TCIAB6029-1G
TCIAB6029-200MG
3-(Benzyloxy)cyclobutanone ≥95.0% (by GC)
3-(Benzyloxy)cyclobutanone
Formel:
C11H12O2 Kogepunkt: 102°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD09755991 CAS-nummer: 30830-27-4 |
Specifikationer Testresultater
Appearance | Colorless to Yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...