(4-Bromophenylethynyl)trimethylsilane

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: ((4-Bromophenyl)ethynyl)trimethylsilane

B6028-1G B6028-5G
TCIAB6028-1GEA 214 DKK
TCIAB6028-1G TCIAB6028-5G
(4-Bromophenylethynyl)trimethylsilane
(4-Bromophenylethynyl)trimethylsilane
Formel: C₁₁H₁₃BrSi
Smeltepunkt: 62°C
MDL Number: MFCD01863556
CAS-nummer: 16116-78-2

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 60.0 to 64.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR