3-Bromo-5-methylaniline ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B6015-25G B6015-5G
TCIAB6015-25GEA 3380 DKK
TCIAB6015-25G TCIAB6015-5G
3-Bromo-5-methylaniline ≥98.0% (by GC, titration analysis)
3-Bromo-5-methylaniline
Formel: C7H8BrN
Kogepunkt: 260°C
Smeltepunkt: 37°C
MDL Number: MFCD11845553
CAS-nummer: 74586-53-1
EINECS: 277-932-1

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR