2-(1-tert-Butoxycarbonylazetidin-3-yl)acetic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B6013-1G B6013-5G
TCIAB6013-1GEA 132 DKK
TCIAB6013-1G TCIAB6013-5G
2-(1-tert-Butoxycarbonylazetidin-3-yl)acetic acid ≥98.0% (by GC, titration analysis)
2-(1-tert-Butoxycarbonylazetidin-3-yl)acetic acid
Formel: C₁₀H₁₇NO₄
MDL Number: MFCD02179028
CAS-nummer: 183062-96-6

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 116.0 to 120.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR