tert-Butyl-3-methyleneazetidine-1-carboxylate ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B6011-1G B6011-5G
TCIAB6011-1GEA 435 DKK
TCIAB6011-1G TCIAB6011-5G
tert-Butyl-3-methyleneazetidine-1-carboxylate ≥95.0% (by GC)
tert-Butyl-3-methyleneazetidine-1-carboxylate
Formel: C₉H₁₅NO₂
MDL Number: MFCD12031229
CAS-nummer: 934664-41-2

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR