1-Brom-3-fluor-2-iodbenzen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B5982-1G B5982-5G
TCIAB5982-1GEA 214 DKK
TCIAB5982-1G TCIAB5982-5G
1-Brom-3-fluor-2-iodbenzen ≥98.0% (by GC)
1-Brom-3-fluor-2-iodbenzen
Formel: C₆H₃BrFI
Molekylvægt: 300.8949 g/mol
Kogepunkt: 109°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD07368675
CAS-nummer: 450412-29-0

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR