2-(tert-Butylthio)ethylamine hydrochloride ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B5979-25G
B5979-5G
TCIAB5979-25GEA
3250
DKK
TCIAB5979-25G
TCIAB5979-5G
2-(tert-Butylthio)ethylamine hydrochloride ≥98.0% (by GC, titration analysis)
2-(tert-Butylthio)ethylamine hydrochloride
Formel:
C₆H₁₅NS·HCl Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00052000 CAS-nummer: 60116-77-0 EINECS: 262-065-3 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 208.0 to 212.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...