1,6-Bis(trichlorosilyl)hexane ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
B5923-25G
B5923-5G
TCIAB5923-25GEA
2610
DKK
TCIAB5923-25G
TCIAB5923-5G
1,6-Bis(trichlorosilyl)hexane ≥95.0% (by GC)
1,6-Bis(trichlorosilyl)hexane
Formel:
C₆H₁₂Cl₆Si₂ Molekylvægt: 353.03 g/mol Kogepunkt: 281°C Flammepunkt: 75°C |
MDL Number:
MFCD00053189 CAS-nummer: 13083-94-8 EINECS: 235-994-7 UN nummer: 2987 ADR: 8 II |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...