4,4'-Bis(trans-4-propylcyclohexyl)biphenyl ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4,4'-Bis(trans-4-propylcyclohexyl)-1,1'-biphenyl

B5881-25G B5881-5G
TCIAB5881-25GEA 2040 DKK
TCIAB5881-25G TCIAB5881-5G
4,4'-Bis(trans-4-propylcyclohexyl)biphenyl ≥98.0% (by GC)
4,4'-Bis(trans-4-propylcyclohexyl)biphenyl
Formel: C₃₀H₄₂
Molekylvægt: 402.67 g/mol
MDL Number: MFCD11053445
CAS-nummer: 85600-56-2

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR