Butyltriethoxysilane ≥94.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B5858-25ML B5858-5ML
TCIAB5858-25MLEA 739 DKK
TCIAB5858-25ML TCIAB5858-5ML
Butyltriethoxysilane ≥94.0% (by GC)
Butyltriethoxysilane
Formel: C₁₀H₂₄O₃Si
Kogepunkt: 193°C
MDL Number: MFCD00138096
CAS-nummer: 4781-99-1

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR