(2R,4R)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Boc-D-cis-Hyp-OMe, N-(tert-Butoxycarbonyl)-cis-4-hydroxy-D-proline methyll ester, 1-tert-Butyl 2-Methyl (2R,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylate, Boc-cis-D-Hyp-OMe, 1-tert-butyl 2-methyl (2R,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate, (2R,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylic acid 1-tert-Butyl 2-Methyl ester, (2R,4R)-1-tert-Butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, N-Boc-cis-4-hydroxy-D-proline methyll ester, N-Boc-cis-4-hydroxy-D-proline methyl ester

B5857-1G B5857-5G
TCIAB5857-1GEA 173 DKK
TCIAB5857-1G TCIAB5857-5G
(2R,4R)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate ≥97.0% (by GC)
(2R,4R)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate
Formel: C₁₁H₁₉NO₅
Molekylvægt: 245.27 g/mol
MDL Number: MFCD00797548
CAS-nummer: 114676-69-6

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 78.0 to 82.0 deg-C
Specific rotation [a]20/D +62.0 to +66.0 deg (C=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR