2-Chlorphenethylbromid ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-(2-Bromoethyl)-2-chlorobenzene, 2-Chlorophenethyl Bromide, 2-(2-Chlorophenyl)ethyl bromide, 2-Chlorophenethyl bromide

B5833-1G B5833-5G
TCIAB5833-1GEA 263 DKK
TCIAB5833-1G TCIAB5833-5G
2-Chlorphenethylbromid ≥98.0% (by GC)
2-Chlorphenethylbromid
Formel: C₈H₈BrCl
Molekylvægt: 219.51 g/mol
Kogepunkt: 112°C
MDL Number: MFCD04117464
CAS-nummer: 16793-91-2

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR