2-Bromobenzaldehyde Dimethyl Acetal ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Bromo-2-(dimethoxymethyl)benzene

B5827-5G B5827-25G
TCIAB5827-5GEA 304 DKK
TCIAB5827-5G TCIAB5827-25G
2-Bromobenzaldehyde Dimethyl Acetal ≥98.0% (by GC)
2-Bromobenzaldehyde Dimethyl Acetal
Formel: C₉H₁₁BrO₂
Molekylvægt: 231.09 g/mol
Kogepunkt: 103°C
MDL Number: MFCD07780683
CAS-nummer: 35849-09-3

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR