4-Brom-2-fluorbenzoylchlorid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
4-Bromo-2-fluorobenzene-1-carbonyl chloride
B5817-25G
B5817-5G
TCIAB5817-25GEA
1990
DKK
TCIAB5817-25G
TCIAB5817-5G
4-Brom-2-fluorbenzoylchlorid ≥98.0% (by GC, titration analysis)
4-Brom-2-fluorbenzoylchlorid
Formel:
C₇H₃BrClFO |
MDL Number:
MFCD03411584 CAS-nummer: 151982-51-3 UN nummer: 3265 ADR: 8,II |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...